accessibility menu, dialog, popup

UserName

CAS RN 185057-50-5

NNH2OO

CAS RN 185057-50-5

IUPAC Name: benzyl 3-aminopyrrolidine-1-carboxylate
Synonyms: benzyl 3-aminopyrrolidine-1-carboxylate
Molecular Weight (g/mol): 220.27
Molecular Formula: C12H16N2O2
InChi Key: FPXJNSKAXZNWMQ-UHFFFAOYNA-N
SMILES: NC1CCN(C1)C(=O)OCC1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results