missing translation for 'onlineSavingsMsg'
Learn More

3-Acetoxy-2-methylbenzoyl chloride, 97%, Thermo Scientific™

Product Code. p-7047332
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11359495 5 g 5g
11369495 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11359495 Supplier Thermo Scientific Alfa Aesar Supplier No. L18720.06

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167678-46-8
Molecular Formula C10H9ClO3
Molecular Weight (g/mol) 212.63
MDL Number MFCD02094037
InChI Key COLDUSGLGQXXEJ-UHFFFAOYSA-N
Synonym 3-acetoxy-2-methylbenzoyl chloride, 3-chlorocarbonyl-2-methylphenyl acetate, 3-acetoxy-2-methylbenzoylchloride, 3-acetyloxy-2-methyl-benzoyl chloride, 3-carbonochloridoyl-2-methylphenyl acetate, acetic acid 3-chlorocarbonyl-2-methylphenyl ester, ambc, acmc-1c4md, 3-acetoxy-o-toluoyl chloride, 3-acetoxy-2-methyl-benzoyl chloride
PubChem CID 7010345
SMILES CC(=O)OC1=CC=CC(C(Cl)=O)=C1C

Specifications

Melting Point 31°C to 35°C
Density 1.25
Quantity 5 g
UN Number UN3261
Beilstein 8054543
Sensitivity Moisture sensitive
IUPAC Name (3-carbonochloridoyl-2-methylphenyl) acetate
Formula Weight 212.63
Percent Purity 97%
Chemical Name or Material 3-Acetoxy-2-methylbenzoyl chloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.