accessibility menu, dialog, popup

UserName

CH3OOCH3OCl

CAS RN 167678-46-8

IUPAC Name: 3-(carbonochloridoyl)-2-methylphenyl acetate
Synonyms: 3-(carbonochloridoyl)-2-methylphenyl acetate
Molecular Weight (g/mol): 212.63
Molecular Formula: C10H9ClO3
InChi Key: COLDUSGLGQXXEJ-UHFFFAOYSA-N
SMILES: CC(=O)OC1=CC=CC(C(Cl)=O)=C1C

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results