missing translation for 'onlineSavingsMsg'
Learn More

3-(9H-Carbazol-9-yl)propanoic Acid, TRC

Product Code. 30390636
Change view
Click to view available options
Quantity:
250 mg
500 mg
1 g
Unit Size:
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30390636 250 mg 250mg
30394599 500 mg -
30404945 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30390636 Supplier Toronto Research Chemicals Supplier No. TRCH956178250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6622-54-4
Molecular Formula C15 H13 N O2
Molecular Weight (g/mol) 239.269
IUPAC Name 3-carbazol-9-ylpropanoic acid
SMILES OC(=O)CCn1c2ccccc2c3ccccc13

Specifications

Quantity 250 mg
InChI Formula InChI=1S/C15H13NO2/c17-15(18)9-10-16-13-7-3-1-5-11(13)12-6-2-4-8-14(12)16/h1-8H,9-10H2,(H,17,18)
Formula Weight 239.095
Chemical Name or Material 3-(9H-Carbazol-9-yl)propanoic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.