CAS RN 6622-54-4
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-(9H-Carbazol-9-yl)propanoic Acid, TRC
CAS: 6622-54-4 Molecular Formula: C15 H13 N O2 Molecular Weight (g/mol): 239.269 IUPAC Name: 3-carbazol-9-ylpropanoic acid SMILES: OC(=O)CCn1c2ccccc2c3ccccc13