missing translation for 'onlineSavingsMsg'
Learn More

3,7-Dimethyl-1-propargylxanthine, TRC

Product Code. 30369719
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30369719 5 mg
30386642 10 mg
30370113 25 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30369719 Supplier Toronto Research Chemicals Supplier No. TRCD4778505MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14114-46-6
Molecular Formula C10 H10 N4 O2
Molecular Weight (g/mol) 218.21
IUPAC Name 3,7-dimethyl-1-prop-2-ynylpurine-2,6-dione
SMILES CN1C(=O)N(CC#C)C(=O)c2c1ncn2C

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C10H10N4O2/c1-4-5-14-9(15)7-8(11-6-12(7)2)13(3)10(14)16/h1,6H,5H2,2-3H3
Formula Weight 218.0804
Chemical Name or Material 3,7-Dimethyl-1-propargylxanthine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.