CAS RN 14114-46-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,7-Dimethyl-1-propargylxanthine, TRC
CAS: 14114-46-6 Molecular Formula: C10 H10 N4 O2 Molecular Weight (g/mol): 218.21 IUPAC Name: 3,7-dimethyl-1-prop-2-ynylpurine-2,6-dione SMILES: CN1C(=O)N(CC#C)C(=O)c2c1ncn2C