missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dimethylphenylmagnesium bromide, 0.5M in 2-MeTHF

Product Code. 15470706
Change view
Click to view available options
Quantity:
100 mL
Unit Size:
100mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15470706 100 mL 100mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15470706 Supplier Thermo Scientific Alfa Aesar Supplier No. H54057.AE

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 34696-73-6 | C8H9BrMg | 209.37 g/mol

3,5-Dimethylphenylmagnesium bromide is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dimethylphenylmagnesium bromide is used as pharmaceutical intermediate.

Solubility
It reacts with water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents and bases.
TRUSTED_SUSTAINABILITY

Specifications

CAS 34696-73-6
Molecular Formula C8H9BrMg
MDL Number MFCD01311497
Quantity 100 mL
UN Number UN2924
Sensitivity Air and moisture sensitive
Synonym 3,5-dimethylphenylmagnesium bromide, xpxqqjmqeswikm-uhfffaoysa-m, 3,5-dimethylphenyl magnesium bromide, 3,5-dimethylphenylmagnesium bromide solution, 3,5-dimethylphenylmagnesium bromide, 0.5m in 2-methf, 3,5-dimethylphenylmagnesium bromide 0.5 m in tetrahydrofuran, 3,5-dimethylphenylmagnesium bromide solution, 0.5 m in thf, grignard reagent
Solubility Information It reacts with water.
InChI Key XPXQQJMQESWIKM-UHFFFAOYSA-M
SMILES CC1=CC([Mg]Br)=CC(C)=C1
IUPAC Name magnesium;1,3-dimethylbenzene-5-ide;bromide
Molecular Weight (g/mol) 209.37
PubChem CID 10965748
Formula Weight 209.37
Concentration or Composition (by Analyte or Components) 0.5M in 2-MeTHF
Chemical Name or Material 3,5-Dimethylphenylmagnesium bromide
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.