missing translation for 'onlineSavingsMsg'
Learn More

3,5-Diiodo-4-hydroxyphenylpropionic Acid, TRC

Product Code. 30386099
Change view
Click to view available options
Quantity:
1 g
2.5 g
5 g
Unit Size:
2.5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30386099 5 g -
30377100 1 g -
30378709 2.5 g 2.5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30386099 Supplier Toronto Research Chemicals Supplier No. TRCD4551835G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13811-11-5
Molecular Formula C9H8I2O3
Molecular Weight (g/mol) 417.97
Synonym 4-Hydroxy-3,5-diiodobenzenepropanoic Acid, 3,5-Diiodophloretic Acid, 3-(4-Hydroxy-3,5-diiodophenyl)propanoic Acid, 3-(4-Hydroxy-3,5-diiodophenyl)propionic Acid, 3,5-Diiodo-phloretic Acid, 4-Hydroxy-3,5-diiodo-benzenepropanoic Acid
SMILES OC(=O)CCc1cc(I)c(O)c(I)c1

Specifications

Quantity 5 g
InChI Formula InChI=1S/C9H8I2O3/c10-6-3-5(1-2-8(12)13)4-7(11)9(6)14/h3-4,14H,1-2H2,(H,12,13)
Formula Weight 417.8563
Chemical Name or Material 3,5-Diiodo-4-hydroxyphenylpropionic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.