CAS RN 13811-11-5
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,5-Diiodo-4-hydroxyphenylpropionic Acid, TRC
CAS: 13811-11-5 Molecular Formula: C9H8I2O3 Molecular Weight (g/mol): 417.97 Synonym: 4-Hydroxy-3,5-diiodobenzenepropanoic Acid,3,5-Diiodophloretic Acid,3-(4-Hydroxy-3,5-diiodophenyl)propanoic Acid,3-(4-Hydroxy-3,5-diiodophenyl)propionic Acid,3,5-Diiodo-phloretic Acid,4-Hydroxy-3,5-diiodo-benzenepropanoic Acid SMILES: OC(=O)CCc1cc(I)c(O)c(I)c1