missing translation for 'onlineSavingsMsg'
Learn More

3',5-Dihydroxy-4',6,7-trimethoxyflavone, 97%

Product Code. p-7046473
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
This item is not returnable. View return policy

Product Code. 11393546

Brand: Thermo Scientific Alfa Aesar L14495.MB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 855-96-9 | C18H16O7 | 344.319 g/mol

3',5-Dihydroxy-4',6,7-trimethoxyflavone is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3′,5-Dihydroxy-4′,6,7-trimethoxyflavone is an important raw material and intermediate used in organic synthesis, pharmaceuticals and agrochemicals.

Solubility
Insoluble in water. Soluble in chloroform and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 855-96-9
Molecular Formula C18H16O7
Molecular Weight (g/mol) 344.319
MDL Number MFCD00016929
InChI Key KLAOKWJLUQKWIF-UHFFFAOYSA-N
Synonym eupatorin, 3',5-dihydroxy-4',6,7-trimethoxyflavone, eupatorine, 5-hydroxy-2-3-hydroxy-4-methoxyphenyl-6,7-dimethoxy-4h-chromen-4-one, unii-3j474av6my, 5-hydroxy-2-3-hydroxy-4-methoxyphenyl-6,7-dimethoxy-4h-1-benzopyran-4-one, 6-methoxyluteolin 4',7-dimethyl ether, 5-hydroxy-2-3-hydroxy-4-methoxyphenyl-6,7-dimethoxychromen-4-one, kinome_3214, spectrum_001037
PubChem CID 97214
IUPAC Name 5-hydroxy-2-(3-hydroxy-4-methoxyphenyl)-6,7-dimethoxychromen-4-one
SMILES COC1=C(C=C(C=C1)C2=CC(=O)C3=C(C(=C(C=C3O2)OC)OC)O)O

Specifications

Melting Point 190°C to 192°C
Quantity 25 mg
Beilstein 1355109
Merck Index 14,3901
Solubility Information Insoluble in water. Soluble in chloroform and methanol.
Formula Weight 344.32
Percent Purity 97%
Chemical Name or Material 3',5-Dihydroxy-4',6,7-trimethoxyflavone

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.