missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorocinnamic acid, 98%, Thermo Scientific™

Product Code. 10155560 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10155560 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10155560 Supplier Thermo Scientific Acros Supplier No. 311590010
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 84315-23-1
CAS Min % 97.5
CAS Max % 100.0
Melting Point 203°C to 206°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C9H6F2O2
Linear Formula F2C6H3CH=CHCO2H
MDL Number MFCD00010321
Quantity 1 g
Synonym 3,5-difluorocinnamic acid, trans-3,5-difluorocinnamic acid, 3,5-difluorocinnamicacid, e-3-3,5-difluorophenyl acrylic acid, 3-3,5-difluorophenyl acrylic acid, 2e-3-3,5-difluorophenyl prop-2-enoic acid, 2e-3-3,5-difluorophenyl acrylic acid, rarechem bk hw 0255, timtec-bb sbb006675, 3-3,5-difluorophenyl propenoic acid
InChI Key MBAWRXICVNIUGY-OWOJBTEDSA-N
SMILES C1=C(C=C(C=C1F)F)C=CC(=O)O
IUPAC Name (E)-3-(3,5-difluorophenyl)prop-2-enoic acid
Molecular Weight (g/mol) 184.14
PubChem CID 5374941
Formula Weight 184.14
Percent Purity 98%
Physical Form Fluffy Shiny Powder or Needles
Chemical Name or Material 3, 5-Difluorocinnamic acid, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.