missing translation for 'onlineSavingsMsg'
Learn More

3,5-Difluorobenzophenone, 98+%, Thermo Scientific™

Product Code. 11438937
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11438937

Brand: Thermo Scientific Alfa Aesar B20554.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 179113-89-4
Molecular Formula C13H8F2O
Molecular Weight (g/mol) 218.20
MDL Number MFCD00061140
InChI Key PNRLNUIMFIMZLI-UHFFFAOYSA-N
Synonym 3,5-difluorobenzophenone, 3,5-difluorophenyl phenyl methanone, 3,5-difluorophenyl-phenylmethanone, 3,5-difluorophenyl phenyl ketone, fr cf evr, acmc-1c2gw, 3,5-difluorobenzophenone, 98+%
PubChem CID 569563
IUPAC Name (3,5-difluorophenyl)-phenylmethanone
SMILES FC1=CC(=CC(F)=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 57°C to 59°C
Boiling Point 116°C to 118°C (5 mmHg)
Quantity 5 g
Beilstein 7919206
Formula Weight 218.2
Percent Purity ≥98%
Chemical Name or Material 3,5-Difluorobenzophenone
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.