accessibility menu, dialog, popup

UserName

CAS RN 179113-89-4

FFO

CAS RN 179113-89-4

IUPAC Name: (3,5-difluorophenyl)(phenyl)methanone
Synonyms: (3,5-difluorophenyl)(phenyl)methanone
Molecular Weight (g/mol): 218.2
Molecular Formula: C13H8F2O
InChi Key: PNRLNUIMFIMZLI-UHFFFAOYSA-N
SMILES: FC1=CC(=CC(F)=C1)C(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results