missing translation for 'onlineSavingsMsg'
Learn More

3,5-Dichloropyridin-4(1H)-one, TRC

Product Code. 30381314
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381314 5 g 5g
30387690 10 g -
30369099 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381314 Supplier Toronto Research Chemicals Supplier No. TRCD4759035G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17228-70-5
Molecular Formula C5H3NOCl2
Molecular Weight (g/mol) 163.99
SMILES ClC1=CNC=C(Cl)C1=O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C5H3Cl2NO/c6-3-1-8-2-4(7)5(3)9/h1-2H,(H,8,9)
Formula Weight 162.9592
Chemical Name or Material 3,5-Dichloropyridin-4(1H)-one
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.