CAS RN 17228-70-5
IUPAC Name:
3,5-dichloro-1,4-dihydropyridin-4-one
Synonyms:
3,5-dichloro-1H-pyridin-4-one
Molecular Weight (g/mol):
163.99
Molecular Formula:
C5H3Cl2NO
InChi Key:
YKCQWIYRLMNGMO-UHFFFAOYSA-N
SMILES:
ClC1=CNC=C(Cl)C1=O
1
–
1
of
1
results
1
3,5-Dichloropyridin-4(1H)-one, TRC
CAS: 17228-70-5 Molecular Formula: C5H3NOCl2 Molecular Weight (g/mol): 163.99 SMILES: ClC1=CNC=C(Cl)C1=O