missing translation for 'onlineSavingsMsg'
Learn More

3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 97%

Product Code. 11448773
Change view
Click to view available options
Quantity:
10 g
50 g
Unit Size:
10g
50g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11448773 50 g 50g
11438773 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11448773 Supplier Thermo Scientific Alfa Aesar Supplier No. A10514.18
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 22014-01-3 | C17H24O3 | 276.38 g/mol

3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans,

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans,

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22014-01-3
Molecular Formula C17H24O3
Molecular Weight (g/mol) 276.38
MDL Number MFCD00017291
InChI Key CTYWXRDQWMRIIM-BQYQJAHWSA-N
Synonym 3-3,5-di-tert-butyl-4-hydroxyphenyl acrylic acid, unii-k7zj1m9j5t, k7zj1m9j5t, 3,5-di-tert-butyl-4-hydroxycinnamic acid, 3,5-di-t-butyl-4-hydroxycinnamic acid, 3,5-di-tert-butyl-4-hydroxycinnamic acid, predominantly trans, 2e-3-3,5-di-tert-butyl-4-hydroxyphenyl prop-2-enoic acid, 3-3,5-di-tert-butyl-4-hydroxyphenyl prop-2-enoic acid, 2e-3-3,5-bis tert-butyl-4-hydroxyphenyl prop-2-enoic acid, 2e-3-3,5-ditert-butyl-4-hydroxyphenyl acrylic acid
PubChem CID 689095
IUPAC Name (E)-3-(3,5-ditert-butyl-4-hydroxyphenyl)prop-2-enoic acid
SMILES CC(C)(C)C1=CC(\C=C\C(O)=O)=CC(=C1O)C(C)(C)C

Specifications

Melting Point ∼195°C (decomposition)
Quantity 50 g
Beilstein 3144594
Solubility Information Soluble in water.
Formula Weight 276.37
Percent Purity 98%
Chemical Name or Material 3,5-Di-tert-butyl-4-hydroxycinnamic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.