missing translation for 'onlineSavingsMsg'
Learn More

3-[[4-(Trifluoromethyl)phenyl]methyl]azetidine Hydrochloride, TRC

Product Code. 30455118
Change view
Click to view available options
Quantity:
250 mg
2.5 g
Unit Size:
2.5g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30455118 2.5 g 2.5g
30455326 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30455118 Supplier Toronto Research Chemicals Supplier No. TRCT7914052.5G
 Restricted Item
Estimated Shipment: 21-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1203685-18-0
Molecular Formula C11H12F3N . HCl
Molecular Weight (g/mol) 251.67
SMILES FC(F)(F)c1ccc(CC2CNC2)cc1

Specifications

Quantity 2.5 g
InChI Formula InChI=1S/C11H12F3N.ClH/c12-11(13,14)10-3-1-8(2-4-10)5-9-6-15-7-9;/h1-4,9,15H,5-7H2;1H
Chemical Name or Material 3-((4-(Trifluoromethyl)phenyl)methyl)azetidine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.