CAS RN 1203685-18-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3-[[4-(Trifluoromethyl)phenyl]methyl]azetidine Hydrochloride, TRC
CAS: 1203685-18-0 Molecular Formula: C11H12F3N . HCl Molecular Weight (g/mol): 251.67 SMILES: FC(F)(F)c1ccc(CC2CNC2)cc1