missing translation for 'onlineSavingsMsg'
Learn More

3-[4-(Trifluoromethoxy)phenyl]acrylic acid, 97%, Thermo Scientific™

Product Code. 10371843
Change view
Click to view available options
Quantity:
1 g
10 g
50 g
Unit Size:
10g
1g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10371843 50 g 50g
10087402 1 g 1g
10757183 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10371843 Supplier Thermo Scientific Maybridge Supplier No. SEW04310EE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 783-13-1
Molecular Formula C10H7F3O3
Molecular Weight (g/mol) 232.158
MDL Number MFCD00066338
InChI Key RNYVTJANWYBGPW-ZZXKWVIFSA-N
Synonym 3-4-trifluoromethoxy phenyl acrylic acid, 4-trifluoromethoxy cinnamic acid, e-3-4-trifluoromethoxy phenyl acrylic acid, e-3-4-trifluoromethoxy phenyl prop-2-enoic acid, 2e-3-4-trifluoromethoxy phenyl prop-2-enoic acid, 3-4-trifluoromethoxyphenyl acrylic acid, trifluoromethoxyphenylacrylicacid, trans-4-trifluoromethoxy cinnamic acid
PubChem CID 735857
IUPAC Name (E)-3-[4-(trifluoromethoxy)phenyl]prop-2-enoic acid
SMILES C1=CC(=CC=C1C=CC(=O)O)OC(F)(F)F

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 50 g
Formula Weight 232.16
Percent Purity 97%
Chemical Name or Material 3-[4-(Trifluoromethoxy)phenyl]acrylic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.