missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinylcarbonyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15439647
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15439647 1 g 1g
15430166 250 mg 250mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15439647 Supplier Thermo Scientific Alfa Aesar Supplier No. H52868.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 723281-55-8
Molecular Formula C11H14BNO4
Molecular Weight (g/mol) 235.046
MDL Number MFCD04115687
InChI Key DRZFURCXDFRZNR-UHFFFAOYSA-N
Synonym 3-morpholine-4-carbonyl phenylboronic acid, n-morpholinyl 3-boronobenzamide, 3-morpholine-4-carbonyl benzeneboronic acid, 3-morpholine-4-carbonyl phenyl boronic acid, 3-morpholin-4-ylcarbonyl phenylboronic acid, boronic acid, 3-4-morpholinylcarbonyl phenyl, acmc-209omm, 4-3-boronobenzoyl morpholine, 3-morpholin-4-carbonyl phenylboronic acid
PubChem CID 3602916
IUPAC Name [3-(morpholine-4-carbonyl)phenyl]boronic acid
SMILES B(C1=CC(=CC=C1)C(=O)N2CCOCC2)(O)O

Specifications

Melting Point 124°C to 130°C
Quantity 1 g
Formula Weight 235.05
Percent Purity 98%
Chemical Name or Material 3-(4-Morpholinylcarbonyl)benzeneboronic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.