accessibility menu, dialog, popup

UserName

CAS RN 723281-55-8

BONOHOOH

CAS RN 723281-55-8

IUPAC Name: [3-(morpholine-4-carbonyl)phenyl]boronic acid
Synonyms: 3-(morpholine-4-carbonyl)phenylboronic acid
Molecular Weight (g/mol): 235.05
Molecular Formula: C11H14BNO4
InChi Key: DRZFURCXDFRZNR-UHFFFAOYSA-N
SMILES: OB(O)C1=CC(=CC=C1)C(=O)N1CCOCC1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results