missing translation for 'onlineSavingsMsg'
Learn More

3-(4-Morpholinyl)propiophenone hydrochloride, 98+%, Thermo Scientific™

Product Code. p-7027151
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11420098 5 g 5g
11430098 25 g 25g
11440098 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11420098 Supplier Thermo Scientific Alfa Aesar Supplier No. B23686.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1020-16-2
Molecular Formula C13H18ClNO2
Molecular Weight (g/mol) 255.742
MDL Number MFCD00035332
InChI Key BEUAFINDWSQCGS-UHFFFAOYSA-N
Synonym 3-morpholinopropiophenone hydrochloride, 3-4-morpholino propiophenone hydrochloride, 3-4-morpholinyl propiophenone hydrochloride, 3-morpholino-1-phenylpropan-1-one hydrochloride, labotest-bb lt00454997, 3-4-morpholinyl-1-phenyl-1-propanone hydrochloride, 3-morpholin-4-yl-1-phenylpropan-1-one hydrochloride, 3-morpholinopropiophenone hcl
PubChem CID 3083670
IUPAC Name 3-morpholin-4-yl-1-phenylpropan-1-one;hydrochloride
SMILES C1COCCN1CCC(=O)C2=CC=CC=C2.Cl

Specifications

Melting Point 180°C to 182°C
Quantity 5 g
Beilstein 3723608
Sensitivity Hygroscopic
Formula Weight 255.76
Percent Purity ≥98%
Chemical Name or Material 3-(4-Morpholinyl)propiophenone hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.