accessibility menu, dialog, popup

UserName

CAS RN 1020-16-2

HClONO

CAS RN 1020-16-2

IUPAC Name: hydrogen 3-(morpholin-4-yl)-1-phenylpropan-1-one chloride
Synonyms: hydrogen 3-(morpholin-4-yl)-1-phenylpropan-1-one chloride
Molecular Weight (g/mol): 255.74
Molecular Formula: C13H18ClNO2
InChi Key: BEUAFINDWSQCGS-UHFFFAOYSA-N
SMILES: [H+].[Cl-].O=C(CCN1CCOCC1)C1=CC=CC=C1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results