missing translation for 'onlineSavingsMsg'
Learn More

3,4-(Methylenedioxy)cinnamic acid, predominantly trans, 99%

Product Code. 11419372
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11419372 25 g 25g
11429372 100 g 100g
11439372 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11419372 Supplier Thermo Scientific Alfa Aesar Supplier No. A10122.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2373-80-0 | C10H8O4 | 192.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2373-80-0
Molecular Formula C10H8O4
Molecular Weight (g/mol) 192.17
MDL Number MFCD00005837
InChI Key QFQYZMGOKIROEC-DUXPYHPUSA-N
Synonym 3,4-methylenedioxycinnamic acid, 3,4-methylenedioxy cinnamic acid, 3-benzo d 1,3 dioxol-5-yl acrylic acid, unii-x8h0wpj08y, 3-1,3-benzodioxol-5-yl acrylic acid, 2e-3-2h-1,3-benzodioxol-5-yl prop-2-enoic acid, methylenedioxycinnamic acid, x8h0wpj08y, 2e-3-1,3-benzodioxol-5-yl acrylic acid, 2e-3-1,3-benzodioxol-5-yl-2-propenoic acid
PubChem CID 643181
ChEBI CHEBI:81482
IUPAC Name (E)-3-(1,3-benzodioxol-5-yl)prop-2-enoic acid
SMILES C1OC2=C(O1)C=C(C=C2)C=CC(=O)O

Specifications

Melting Point ∼243°C (decomposition)
Quantity 25 g
Beilstein 84599
Formula Weight 192.17
Percent Purity 0.99
Chemical Name or Material 3,4- (Methylenedioxy)cinnamic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.