missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3-(4-Methoxyphenyl)-2-phenylpropionic acid

Product Code. p-7033164
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11439320 250 mg 250mg
11449320 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11439320 Supplier Alfa Aesar™ Supplier No. H50213.MD

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 4314-68-5
Molecular Formula C16H16O3
Molecular Weight (g/mol) 256.301
MDL Number MFCD00046532
InChI Key GIROORGDVPRQFZ-UHFFFAOYSA-N
Synonym 3-4-methoxyphenyl-2-phenylpropanoic acid, 3-4-methoxy-phenyl-2-phenyl-propionic acid, 3-4-methoxyphenyl-2-phenylpropionic acid, chembl83949, acmc-20anjp, benzenepropanoic acid,4-methoxy-a-phenyl, 4-methoxy-alpha-phenylbenzenepropanoic acid, 3-4-methoxyphenyl-2-phenyl-propanoic acid, 3-p-methoxyphenyl-2-phenylpropionic acid
PubChem CID 97739
IUPAC Name 3-(4-methoxyphenyl)-2-phenylpropanoic acid
SMILES COC1=CC=C(C=C1)CC(C2=CC=CC=C2)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 256.3
Chemical Name or Material 3-(4-Methoxyphenyl)-2-phenylpropionic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.