CAS RN 4314-68-5
IUPAC Name:
3-(4-methoxyphenyl)-2-phenylpropanoic acid
Synonyms:
3-(4-methoxyphenyl)-2-phenylpropanoic acid
Molecular Weight (g/mol):
256.3
Molecular Formula:
C16H16O3
InChi Key:
GIROORGDVPRQFZ-UHFFFAOYNA-N
SMILES:
COC1=CC=C(CC(C(O)=O)C2=CC=CC=C2)C=C1
1
–
1
of
1
results
1
Alfa Aesar™ 3-(4-Methoxyphenyl)-2-phenylpropionic acid
CAS: 4314-68-5 Molecular Formula: C16H16O3 Molecular Weight (g/mol): 256.301 MDL Number: MFCD00046532 InChI Key: GIROORGDVPRQFZ-UHFFFAOYSA-N Synonym: 3-4-methoxyphenyl-2-phenylpropanoic acid,3-4-methoxy-phenyl-2-phenyl-propionic acid,3-4-methoxyphenyl-2-phenylpropionic acid,chembl83949,acmc-20anjp,benzenepropanoic acid,4-methoxy-a-phenyl,4-methoxy-alpha-phenylbenzenepropanoic acid,3-4-methoxyphenyl-2-phenyl-propanoic acid,3-p-methoxyphenyl-2-phenylpropionic acid PubChem CID: 97739 IUPAC Name: 3-(4-methoxyphenyl)-2-phenylpropanoic acid SMILES: COC1=CC=C(C=C1)CC(C2=CC=CC=C2)C(=O)O