missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethyl-3-cyclopentene-1-carboxylic Acid, TRC

Product Code. 30381216
Change view
Click to view available options
Quantity:
2.5 mg
10 mg
25 mg
Unit Size:
10mg
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30381216 25 mg -
30380472 2.5 mg 2.5mg
30372516 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30381216 Supplier Toronto Research Chemicals Supplier No. TRCD61524525MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 768-38-7
Molecular Formula C8H12O2
Molecular Weight (g/mol) 140.18
Synonym Rosiglitazone Related Compound 1
IUPAC Name 3,4-dimethylcyclopent-3-ene-1-carboxylic acid
SMILES CC1=C(C)CC(C1)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C8H12O2/c1-5-3-7(8(9)10)4-6(5)2/h7H,3-4H2,1-2H3,(H,9,10)
Formula Weight 140.0837
Chemical Name or Material 3,4-Dimethyl-3-cyclopentene-1-carboxylic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.