CAS RN 768-38-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,4-Dimethyl-3-cyclopentene-1-carboxylic Acid, TRC
CAS: 768-38-7 Molecular Formula: C8H12O2 Molecular Weight (g/mol): 140.18 Synonym: Rosiglitazone Related Compound 1 IUPAC Name: 3,4-dimethylcyclopent-3-ene-1-carboxylic acid SMILES: CC1=C(C)CC(C1)C(=O)O