missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,4-Dimethoxycinnamic acid, predominantly trans, 99%, Thermo Scientific™

Product Code. 11454153
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11454153 25 g 25g
11464153 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11454153 Supplier Alfa Aesar™ Supplier No. A10356.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as important components in flavors, perfumes, synthetic indigo and pharmaceuticals.

Solubility
Solubility in hot methanol, very faint turbidity. Soluble in dichloromethane, chloroform.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2316-26-9
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.213
MDL Number MFCD00004387
InChI Key HJBWJAPEBGSQPR-GQCTYLIASA-N
Synonym 3,4-dimethoxycinnamic acid, dimethylcaffeic acid, caffeic acid dimethyl ether, 2e-3-3,4-dimethoxyphenyl prop-2-enoic acid, dimethyl caffeic acid, e-3-3,4-dimethoxyphenyl acrylic acid, e-3,4-dimethoxycinnamic acid, unii-bvz841pvjl, 2e-3-3,4-dimethoxyphenyl acrylic acid, 3-3,4-dimethoxyphenyl acrylic acid
PubChem CID 717531
ChEBI CHEBI:86549
IUPAC Name (E)-3-(3,4-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=C(C=C(C=C1)C=CC(=O)O)OC

Specifications

Melting Point 179°C to 183°C
Quantity 25 g
Beilstein 1533435
Formula Weight 208.21
Percent Purity 99%
Chemical Name or Material 3,4-Dimethoxycinnamic acid, predominantly trans
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.