missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxycinnamic acid, 99%, predominantly trans isomer

Product Code. 10366080 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 10366080

Brand: Thermo Scientific undefined

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 2316-26-9 | C11H12O4 | 208.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2316-26-9
Molecular Formula C11H12O4
Molecular Weight (g/mol) 208.21
MDL Number MFCD00004387
InChI Key HJBWJAPEBGSQPR-GQCTYLIASA-N
Synonym 3,4-dimethoxycinnamic acid, dimethylcaffeic acid, caffeic acid dimethyl ether, 2e-3-3,4-dimethoxyphenyl prop-2-enoic acid, dimethyl caffeic acid, e-3-3,4-dimethoxyphenyl acrylic acid, e-3,4-dimethoxycinnamic acid, unii-bvz841pvjl, 2e-3-3,4-dimethoxyphenyl acrylic acid, 3-3,4-dimethoxyphenyl acrylic acid
PubChem CID 717531
ChEBI CHEBI:86549
IUPAC Name (E)-3-(3,4-dimethoxyphenyl)prop-2-enoic acid
SMILES COC1=C(C=C(C=C1)C=CC(=O)O)OC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 181°C to 184°C
Color Yellow to Beige
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (CH3O)2C6H3CH=CHCO2H
Quantity 100 g
Beilstein 10, 437
Formula Weight 208.21
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3, 4-Dimethoxycinnamic acid, 99%
Show More Show Less
compliance-icons
Product Identifier
  • 3,4-Dimethoxycinnamic acid
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.