missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxybenzaldehyde-d6, TRC

Product Code. 30381100
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30381100 100 mg
30378924 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30381100 Supplier Toronto Research Chemicals Supplier No. TRCD462282100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1162658-05-0
Molecular Formula C9H4D6O3
Molecular Weight (g/mol) 172.21
Synonym 3,4-Dimethoxybenzenecarbonal-d6, 4-O-Methylvanillin-d6, Methylvanillin-d6, NSC 24521-d6, NSC 8500-d6, Protocatechualdehyde Dimethyl Ether-d6, Protocatechuic Aldehyde Dimethyl Ether-d6, Vanillin Methyl Ether-d6, Veratral-d6, Veratric Aldehyde-d6, Veratrum Aldehyde-d6, Veratryl Aldehyde-d6, USP Verapamil Related Compound E-d6
SMILES [2H]C([2H])([2H])OC1=C(OC([2H])([2H])[2H])C=C(C=O)C=C1

Specifications

Quantity 100 mg
InChI Formula InChI=1S/C9H10O3/c1-11-8-4-3-7(6-10)5-9(8)12-2/h3-6H,1-2H3/i1D3,2D3
Chemical Name or Material 3,4-Dimethoxybenzaldehyde-d6
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.