CAS RN 1162658-05-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
3,4-Dimethoxybenzaldehyde-d6, TRC
CAS: 1162658-05-0 Molecular Formula: C9H4D6O3 Molecular Weight (g/mol): 172.21 Synonym: 3,4-Dimethoxybenzenecarbonal-d6,4-O-Methylvanillin-d6,Methylvanillin-d6,NSC 24521-d6,NSC 8500-d6,Protocatechualdehyde Dimethyl Ether-d6,Protocatechuic Aldehyde Dimethyl Ether-d6,Vanillin Methyl Ether-d6,Veratral-d6,Veratric Aldehyde-d6,Veratrum Aldehyde-d6,Veratryl Aldehyde-d6,USP Verapamil Related Compound E-d6 SMILES: [2H]C([2H])([2H])OC1=C(OC([2H])([2H])[2H])C=C(C=O)C=C1