missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxy-N,N-dimethylbenzeneethanamine Hydrochloride, TRC

Product Code. 30382727
Change view
Click to view available options
Quantity:
50 mg
250 mg
500 mg
Unit Size:
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30382727 500 mg 500mg
30379750 250 mg -
30372190 50 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30382727 Supplier Toronto Research Chemicals Supplier No. TRCD476830500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51012-67-0
Molecular Formula C12 H19 N O2 . Cl H
Molecular Weight (g/mol) 245.75
Synonym 2-(3,4-Dimethoxyphenyl)-N,N-dimethylethan-amine Hydrochloride, Verapamil Hydrochloride Imp. C (EP) as Hydrochloride
IUPAC Name 2-(3,4-dimethoxyphenyl)-N,N-dimethylethanamine;hydrochloride
SMILES Cl.COc1ccc(CCN(C)C)cc1OC

Specifications

Quantity 500 mg
InChI Formula InChI=1S/C12H19NO2.ClH/c1-13(2)8-7-10-5-6-11(14-3)12(9-10)15-4;/h5-6,9H,7-8H2,1-4H3;1H
Formula Weight 245.1183
Chemical Name or Material 2-(3,4-Dimethoxyphenyl)-N,N-dimethylethan-amine Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.