CAS RN 51012-67-0
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
2
of
2
results
1
Verapamil EP Impurity C (as Hydrochloride), Mikromol™
Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.
2
3,4-Dimethoxy-N,N-dimethylbenzeneethanamine Hydrochloride, TRC
CAS: 51012-67-0 Molecular Formula: C12 H19 N O2 . Cl H Molecular Weight (g/mol): 245.75 Synonym: 2-(3,4-Dimethoxyphenyl)-N,N-dimethylethan-amine Hydrochloride,Verapamil Hydrochloride Imp. C (EP) as Hydrochloride IUPAC Name: 2-(3,4-dimethoxyphenyl)-N,N-dimethylethanamine;hydrochloride SMILES: Cl.COc1ccc(CCN(C)C)cc1OC