missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dimethoxy[7,8,9,-13C3]-cinnamic Acid, TRC

Product Code. 30380564
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380564 25 mg
30385590 2.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380564 Supplier Toronto Research Chemicals Supplier No. TRCD46066725MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1185241-38-6
Molecular Formula 13C3 C8 H12 O4
Molecular Weight (g/mol) 211.19
Synonym 3-(3,4-Dimethoxyphenyl)-2-propenoic-1,2,3-13C3 acid, 3,4-Di-O-methylcaffeic-13C3 acid, 3,4-Dimethoxycinnamic-13C3 acid, 3-(3,4-Dimethoxyphenyl)acrylic-13C3 acid, Caffeic acid-13C3 dimethyl ether, Methylferulic acid-13C3, 3,4-Dimethoxycinnamic acid-13C3, 3,4-Dimethoxycinnamic acid 13C3 (propenoate-13C3), 3,4-Dimethoxy[7,8,9-13C3]cinnamic acid
IUPAC Name (E)-3-(3,4-dimethoxyphenyl)(1,2,3-^{13}C_{3})prop-2-enoic acid
SMILES COc1ccc(\[13CH]=[13CH]\[13C](=O)O)cc1OC

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C11H12O4/c1-14-9-5-3-8(4-6-11(12)13)7-10(9)15-2/h3-7H,1-2H3,(H,12,13)/b6-4+/i4+1,6+1,11+1
Formula Weight 211.0836
Chemical Name or Material 3,4-Dimethoxy[7,8,9,-13C3]-cinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.