missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ 3,4'-Diisopropylbiphenyl, 94%

Product Code. 11347199
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11347199 100 g 100g
11327199 5 g 5g
11337199 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11347199 Supplier Alfa Aesar™ Supplier No. H27393.22
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 61434-46-6
Molecular Formula C18H22
Molecular Weight (g/mol) 238.37
MDL Number MFCD00043539
InChI Key LHNUPUGVRFQTLK-UHFFFAOYSA-N
Synonym 3,4'-diisopropylbiphenyl, 3,4'-di-iso-propylbiphenyl, 3,4'-diisopropyl-1,1'-biphenyl, unii-gf449ran44, 1,1'-biphenyl,3,4'-bis 1-methylethyl, 1-methylethyl-3-4-methylethyl phenyl benzene, 1-isopropyl-3-4-isopropylphenyl benzene, 3,4-di-iso-propylbiphenyl, 1,1'-biphenyl, 3,4'-diisopropyl, 3,4'-diisopropyl-1,1'-biphenyl #
PubChem CID 123379
IUPAC Name 1-propan-2-yl-3-(4-propan-2-ylphenyl)benzene
SMILES CC(C)C1=CC=C(C=C1)C1=CC=CC(=C1)C(C)C

Specifications

Quantity 100 g
Formula Weight 238.37
Percent Purity 94%
Chemical Name or Material 3,4'-Diisopropylbiphenyl
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.