accessibility menu, dialog, popup

UserName

CAS RN 61434-46-6

H3CCH3CH3H3C

CAS RN 61434-46-6

IUPAC Name: 3,4'-bis(propan-2-yl)-1,1'-biphenyl
Synonyms: 3,4'-diisopropyl-1,1'-biphenyl
Molecular Weight (g/mol): 238.37
Molecular Formula: C18H22
InChi Key: LHNUPUGVRFQTLK-UHFFFAOYSA-N
SMILES: CC(C)C1=CC=C(C=C1)C1=CC=CC(=C1)C(C)C
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results