missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dihydro-2H-1,5-benzodioxepin, 98%, Thermo Scientific™

Product Code. 11337159
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337159 25 g 25g
11317159 1 g 1g
11327159 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11337159 Supplier Thermo Scientific Alfa Aesar Supplier No. H27374.14

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7216-18-4
Molecular Formula C9H10O2
Molecular Weight (g/mol) 150.18
MDL Number MFCD01333457
InChI Key CBXMULHQEVXJDI-UHFFFAOYSA-N
Synonym 3,4-dihydro-2h-benzo b 1,4 dioxepine, 2h-1,5-benzodioxepin, 3,4-dihydro, pyrocatechol trimethylene ether, 3,4-dihydro-2h-1,4-benzodioxepin, 3,4-dihydro-2h-1,5-benzodioxepin, 2h,3h,4h-benzo b 1,4-dioxepin, 1,2-trimethylenedioxybenzene, 1,5-benzodioxepane, 5-19-01-00450 beilstein handbook reference, #
PubChem CID 81635
IUPAC Name 3,4-dihydro-2H-1,5-benzodioxepine
SMILES C1COC2=CC=CC=C2OC1

Specifications

Refractive Index 1.545
Quantity 25 g
Solubility Information Slightly soluble in water.
Formula Weight 150.18
Percent Purity 98%
Chemical Name or Material 3,4-Dihydro-2H-1,5-benzodioxepin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.