CAS RN 7216-18-4
IUPAC Name:
3,4-dihydro-2H-1,5-benzodioxepine
Synonyms:
3,4-dihydro-2H-1,5-benzodioxepine
Molecular Weight (g/mol):
150.18
Molecular Formula:
C9H10O2
InChi Key:
CBXMULHQEVXJDI-UHFFFAOYSA-N
SMILES:
C1COC2=CC=CC=C2OC1
1
–
1
of
1
results
1
3,4-Dihydro-2H-1,5-benzodioxepin, 98%, Thermo Scientific™
CAS: 7216-18-4 Molecular Formula: C9H10O2 Molecular Weight (g/mol): 150.18 MDL Number: MFCD01333457 InChI Key: CBXMULHQEVXJDI-UHFFFAOYSA-N Synonym: 3,4-dihydro-2h-benzo b 1,4 dioxepine,2h-1,5-benzodioxepin, 3,4-dihydro,pyrocatechol trimethylene ether,3,4-dihydro-2h-1,4-benzodioxepin,3,4-dihydro-2h-1,5-benzodioxepin,2h,3h,4h-benzo b 1,4-dioxepin,1,2-trimethylenedioxybenzene,1,5-benzodioxepane,5-19-01-00450 beilstein handbook reference,# PubChem CID: 81635 IUPAC Name: 3,4-dihydro-2H-1,5-benzodioxepine SMILES: C1COC2=CC=CC=C2OC1