missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dichlorocinnamic acid, 97%

Product Code. 11456986
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11456986 5 g 5g
11466986 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11456986 Supplier Thermo Scientific Alfa Aesar Supplier No. B25308.06
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1202-39-7 | C9H6Cl2O2 | 217.045 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1202-39-7
Molecular Formula C9H6Cl2O2
Molecular Weight (g/mol) 217.045
MDL Number MFCD00004385
InChI Key RRLUFPHCTSFKNR-DUXPYHPUSA-N
Synonym 3,4-dichlorocinnamic acid, e-3-3,4-dichlorophenyl acrylic acid, 3',4'-dichlorocinnamic acid, 2-propenoic acid, 3-3,4-dichlorophenyl, cinnamic acid, 3,4-dichloro, 3-3,4-dichlorophenyl acrylic acid, 2e-3-3,4-dichlorophenyl prop-2-enoic acid, 3-3,4-dichlorophenyl-2-propenoic acid, e-3-3,4-dichlorophenyl prop-2-enoic acid, 2e-3-3,4-dichlorophenyl acrylic acid
PubChem CID 688027
IUPAC Name (E)-3-(3,4-dichlorophenyl)prop-2-enoic acid
SMILES C1=CC(=C(C=C1C=CC(=O)O)Cl)Cl

Specifications

Melting Point 218°C to 220°C
Quantity 5 g
Beilstein 1872129
Formula Weight 217.05
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorocinnamic acid

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.