missing translation for 'onlineSavingsMsg'
Learn More

3,4-Dibenzyloxybenzaldehyde, 99%

Product Code. 10240803 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10240803 5 g Glass bottle 5g
10518143 1 g Glass bottle 1g
10005963 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10240803 Supplier Thermo Scientific Acros Supplier No. 112640050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5447-02-9 | C21H18O3 | 318.36 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5447-02-9
Molecular Formula C21H18O3
Molecular Weight (g/mol) 318.36
MDL Number MFCD00004776
InChI Key XDDLXZHBWVFPRG-UHFFFAOYSA-N
Synonym 3,4-dibenzyloxybenzaldehyde, 3,4-bis benzyloxy benzaldehyde, 3,4-dibenzyloxy benzaldehyde, 3,4-bis phenylmethoxy benzaldehyde, xddlxzhbwvfprg-uhfffaoysa-n, 3.4-dibenzyloxybenzaldehyde, 3,4-dibenzyloxy-benzaldehyde, 3,4-dibenzyl-oxybenzaldehyde, benzaldehyde, 3,4-dibenzyloxy, xddlxzhbwvfprg-uhfffaoysa
PubChem CID 79526

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 88°C to 92°C
Color White to Yellow
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula (C6H5CH2O)2C6H3CHO
Quantity 5 g
Formula Weight 318.36
Percent Purity 99%
Physical Form Powder
Chemical Name or Material 3, 4-Dibenzyloxybenzaldehyde
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.