CAS RN 5447-02-9
CAS RN 5447-02-9
IUPAC Name:
3,4-bis(benzyloxy)benzaldehyde
Synonyms:
3,4-bis(benzyloxy)benzaldehyde
Molecular Weight (g/mol):
318.37
Molecular Formula:
C21H18O3
InChi Key:
XDDLXZHBWVFPRG-UHFFFAOYSA-N
SMILES:
O=CC1=CC=C(OCC2=CC=CC=C2)C(OCC2=CC=CC=C2)=C1
1
–
2
of
2
results
1
3,4-Dibenzyloxybenzaldehyde, 99%
CAS: 5447-02-9 Molecular Formula: C21H18O3 Molecular Weight (g/mol): 318.36 MDL Number: MFCD00004776 InChI Key: XDDLXZHBWVFPRG-UHFFFAOYSA-N Synonym: 3,4-dibenzyloxybenzaldehyde,3,4-bis benzyloxy benzaldehyde,3,4-dibenzyloxy benzaldehyde,3,4-bis phenylmethoxy benzaldehyde,xddlxzhbwvfprg-uhfffaoysa-n,3.4-dibenzyloxybenzaldehyde,3,4-dibenzyloxy-benzaldehyde,3,4-dibenzyl-oxybenzaldehyde,benzaldehyde, 3,4-dibenzyloxy,xddlxzhbwvfprg-uhfffaoysa PubChem CID: 79526
2
3,4-Dibenzyloxybenzaldehyde, TRC
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.