missing translation for 'onlineSavingsMsg'
Learn More

[3,4]-Dibenzo-21-crown-7, 98%

Product Code. 10167933 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10167933 5 g 5g
10301324 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10167933 Supplier Thermo Scientific Supplier No. 278160050
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 14098-41-0 | C22H28O7 | 404.45 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chelation/Complexation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14098-41-0
Molecular Formula C22H28O7
Molecular Weight (g/mol) 404.45
MDL Number MFCD00068646
InChI Key JKCQOMAQPUYHPL-UHFFFAOYSA-N
Synonym dibenzo-21-crown-7, dibenzo-21-crown 7-ether, 6,7,9,10,12,13,20,21,23,24-decahydrodibenzo b,k 1,4,7,10,13,16,19 heptaoxacyclohenicosine, dibenzo-21-crown7-ether, dibenzo-21-crown, 2,3,14,15-dibenzo-1,4,7,10,13,16,19-heptaoxacycloheneicosa-2,14-diene, 2,5,8,15,18,21,24-heptaoxatricyclo 23.4.0.0?, 1 ? nonacosa-1 29 ,9,11,13,25,27-hexaene, 6,7,9,10,12,13,20,21,23,24-decahydrodibenzo b,k 1,4,7,10,13,16,19 heptaoxacyclohenicosine #, dibenzo b,k 1,4,7,10,13,16,19 heptaoxacycloheneicosin,6,7,9,10,12,13,20,21,23,24-decahydro
PubChem CID 585932
SMILES C1COCCOC2=CC=CC=C2OCCOCCOC3=CC=CC=C3OCCO1

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 104.0°C to 108.0°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Quantity 5 g
Formula Weight 404.45
Percent Purity 98%
Physical Form Fluffy Amorphous Powder
Chemical Name or Material [3,4]-Dibenzo-21-crown-7, 98%
Show More Show Less
compliance-icons
Product Identifier
  • ¢3,4!-Dibenzo-21-crown-7
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H315-Causes skin irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.