accessibility menu, dialog, popup

UserName

OOOOOOO

CAS RN 14098-41-0

IUPAC Name: 2,5,8,15,18,21,24-heptaoxatricyclo[23.4.0.0⁹,¹⁴]nonacosa-1(29),9,11,13,25,27-hexaene
Synonyms: 2,5,8,15,18,21,24-heptaoxatricyclo[23.4.0.0⁹,¹⁴]nonacosa-1(29),9,11,13,25,27-hexaene
Molecular Weight (g/mol): 404.46
Molecular Formula: C22H28O7
InChi Key: JKCQOMAQPUYHPL-UHFFFAOYSA-N
SMILES: C1COCCOC2=CC=CC=C2OCCOCCOC2=CC=CC=C2OCCO1
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (2)
  • (1)
  • (1)