missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trimethoxycinnamic acid, predominantly trans, 99%

Product Code. 11485103
Change view
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11485103 100 g 100g
11475103 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11485103 Supplier Thermo Scientific Alfa Aesar Supplier No. A12767.22
 Restricted Item
Estimated Shipment: 24-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 90-50-6 | C12H14O5 | 238.239 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-50-6
Molecular Formula C12H14O5
Molecular Weight (g/mol) 238.239
MDL Number MFCD00004388
InChI Key YTFVRYKNXDADBI-SNAWJCMRSA-N
Synonym 3,4,5-trimethoxycinnamic acid, o-methylsinapic acid, 3,4,5-trimethoxyphenylacrylic acid, 3-3,4,5-trimethoxyphenyl acrylic acid, 3-3,4,5-trimethoxyphenyl-2-propenoic acid, 3,4,5-trimethoxy cinnamic acid, sinapic acid methyl ether, e-3-3,4,5-trimethoxyphenyl acrylic acid, cinnamic acid, 3,4,5-trimethoxy, 2e-3-3,4,5-trimethoxyphenyl acrylic acid
PubChem CID 735755
IUPAC Name (E)-3-(3,4,5-trimethoxyphenyl)prop-2-enoic acid
SMILES COC1=CC(=CC(=C1OC)OC)C=CC(=O)O

Specifications

Melting Point 126°C to 128°C
Quantity 100 g
Beilstein 1537834
Formula Weight 238.24
Percent Purity 99%
Chemical Name or Material 3,4,5-Trimethoxycinnamic acid, predominantly trans

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.