missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trihydroxycinnamic Acid, TRC

Product Code. 30457184
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
100mg
1g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30457184 1 g 1g
30450670 500 mg 500mg
30454454 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30457184 Supplier Toronto Research Chemicals Supplier No. TRCT8967731G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6093-59-0
Molecular Formula C9H8O5
Molecular Weight (g/mol) 196.16
Synonym NSC 153688, 3-(3,4,5-Trihydroxyphenyl)-2-propenoic Acid
IUPAC Name (E)-3-(3,4,5-trihydroxyphenyl)prop-2-enoic acid
SMILES OC(=O)\C=C\c1cc(O)c(O)c(O)c1

Specifications

Quantity 1 g
InChI Formula InChI=1S/C9H8O5/c10-6-3-5(1-2-8(12)13)4-7(11)9(6)14/h1-4,10-11,14H,(H,12,13)/b2-1+
Formula Weight 196.0372
Chemical Name or Material 3,4,5-Trihydroxycinnamic Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.