missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorocinnamic acid, 97+%, Thermo Scientific™

Product Code. 11466746
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11466746 1 g 1g
11476746 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11466746 Supplier Thermo Scientific Alfa Aesar Supplier No. B23894.03

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 152152-19-7
Molecular Formula C9H4F3O2
Molecular Weight (g/mol) 201.13
MDL Number MFCD00083529
InChI Key PHIWFMZBVZFEQJ-OWOJBTEDSA-M
Synonym 3,4,5-trifluorocinnamic acid, 2e-3-3,4,5-trifluorophenyl prop-2-enoic acid, 3-3,4,5-trifluoro-phenyl-acrylic acid, 3,4,5-trifluorocinnamicacid, pubchem4351, rarechem bk hw 0056, trans-3,4,5-trifluorocinnamic acid, e-3-3,4,5-trifluorophenyl acrylic acid, e-3-3,4,5-trifluorophenyl prop-2-enoic acid, 2-propenoic acid, 3-3,4,5-trifluorophenyl-, 2e
PubChem CID 5708779
IUPAC Name (E)-3-(3,4,5-trifluorophenyl)prop-2-enoic acid
SMILES [O-]C(=O)\C=C\C1=CC(F)=C(F)C(F)=C1

Specifications

Melting Point 195°C to 199°C
Quantity 1 g
Formula Weight 202.13
Percent Purity ≥97%
Chemical Name or Material 3,4,5-Trifluorocinnamic acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.