Learn More
3',4',5,7-Tetrahydroxyflavone, 97%
Description
3',4',5,7-Tetrahydroxyflavone is used as an apoptosis inducer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3′,4′,5,7-Tetrahydroxyflavone is used as an apoptosis inducer.
Solubility
Soluble in aqueous alkaline solutions (1.4 mg/ml), ethanol (∼5 mg/ml), dimethyl sulfoxide (7 mg/ml), 1eq. Sodium hydroxide (5 mM), dimethylformamide (∼20 mg/ml), water (1 mg/ml) at 25°C and methanol.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.
Specifications
Specifications
| Melting Point | >300°C |
| Quantity | 500 mg |
| Beilstein | 292084 |
| Merck Index | 14,5614 |
| Solubility Information | Soluble in aqueous alkaline solutions (1.4mg/ml),ethanol (∽5mg/ml),dimethyl sulfoxide (7mg/ml),1eq. Sodium hydroxide (5 mM),dimethylformamide (∽20mg/ml),water (1mg/ml) at 25°C and methanol. |
| Formula Weight | 286.24 |
| Percent Purity | 97% |
| Chemical Name or Material | 3',4',5,7-Tetrahydroxyflavone |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.