missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3',4',5,7-Tetrahydroxyflavone, 97%, Thermo Scientific Chemicals
34.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
100 mg
500 mg
Description
3',4',5,7-Tetrahydroxyflavone is used as an apoptosis inducer.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
3′,4′,5,7-Tetrahydroxyflavone is used as an apoptosis inducer.
Solubility
Soluble in aqueous alkaline solutions (1.4 mg/ml), ethanol (∼5 mg/ml), dimethyl sulfoxide (7 mg/ml), 1eq. Sodium hydroxide (5 mM), dimethylformamide (∼20 mg/ml), water (1 mg/ml) at 25°C and methanol.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.
3′,4′,5,7-Tetrahydroxyflavone is used as an apoptosis inducer.
Solubility
Soluble in aqueous alkaline solutions (1.4 mg/ml), ethanol (∼5 mg/ml), dimethyl sulfoxide (7 mg/ml), 1eq. Sodium hydroxide (5 mM), dimethylformamide (∼20 mg/ml), water (1 mg/ml) at 25°C and methanol.
Notes
Incompatible with strong oxidizing agents. Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Recommended storage temperature is 2 - 8°C.
Specifications
Melting Point | >300°C |
Quantity | 100 mg |
Beilstein | 292084 |
Merck Index | 14,5614 |
Solubility Information | Soluble in aqueous alkaline solutions (1.4mg/ml),ethanol (∽5mg/ml),dimethyl sulfoxide (7mg/ml),1eq. Sodium hydroxide (5 mM),dimethylformamide (∽20mg/ml),water (1mg/ml) at 25°C and methanol. |
Formula Weight | 286.24 |
Percent Purity | 97% |
Chemical Name or Material | 3',4',5,7-Tetrahydroxyflavone |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'